missing translation for 'onlineSavingsMsg'
Learn More

Norell™ Ultra Precision Borosilicate Glass NMR Sample Tubes

For auto-sampling systems and high-throughput NMR

Brand:  Norell™ S-5-200-8

 View more versions of this product

Product Code. 12830036

  • 57.10 € / Pack of 5
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Specifically designed for high-resolution NMR, tubes in the Select Series™ are manufactured from ASTM Type 1 Class A borosilicate glass
  • Whether spinning or non-spinning, doing structure determination of small molecules or determining the 3D structure of a membrane protein, the Norell™ Select Series™ of NMR tubes offers unsurpassed performance, reproducibility and reliability compared to other brands
  • You should see an immediate improvement in resolution when calculating i-coupling constants or acquiring data close to your spectrophotometer noise level, resulting in improved peak symmetry and shimmering times
  • In turn this leads to shorter analysis times and improved sample throughput
  • There are also no worries with freeze/thaw degassing or high temperature studies, these tubes can tolerate thermal shock up to 200°C
  • All tubes have polished ends making them less susceptible to breaking
  • Colored end caps are included
Specifications

Specifications

0.035 mm
203 mm
NMR Sample Tube
4.97 mm
Caps
0.0100 mm
Borosilicate Glass
5 Pack
4.2 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Norell™ Ultra Precision Borosilicate Glass NMR Sample Tubes > Camber: 0.035 mm; Concentricity: 0.0100 mm; Length: 203 mm; Inner Diameter: 4.2 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.