Learn More
Norell™ Ultra Precision Borosilicate Glass NMR Sample Tubes
For auto-sampling systems and high-throughput NMR
57.10 € - 181.00 €
Specifications
Material | Borosilicate Glass |
---|---|
Product Type | NMR Sample Tube |
Packaging | 5 Pack |
Includes | Caps |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Camber (Metric) | Concentricity (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Camber (Metric) | Concentricity (Metric) | Price | Quantity & Availability | |||
12880036
|
Norell™
S-3-400-7 |
178 mm | 2.41 mm | 0.019 mm | 0.0070 mm |
114.00 €
Pack of 5 |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||
12860036
|
Norell™
S-3-500-7 |
178 mm | 2.41 mm | 0.013 mm | 0.0050 mm |
137.00 €
Pack of 5 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
12840036
|
Norell™
S-3-600-7 |
178 mm | 2.41 mm | 0.006 mm | 0.0040 mm |
158.00 €
Pack of 5 Qty Price Break Available |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
12830046
|
Norell™
S-3-200-8 |
203 mm | 2.41 mm | 0.038 mm | 0.0100 mm |
59.35 €
Pack of 5 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
12810046
|
Norell™
S-3-300-8 |
203 mm | 2.41 mm | 0.025 mm | 0.0070 mm |
102.00 €
Pack of 5 Qty Price Break Available |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
12890036
|
Norell™
S-3-400-8 |
203 mm | 2.41 mm | 0.019 mm | 0.0070 mm |
132.00 €
Pack of 5 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
12870036
|
Norell™
S-3-500-8 |
203 mm | 2.41 mm | 0.013 mm | 0.0050 mm |
156.00 €
Pack of 5 Qty Price Break Available |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
12850036
|
Norell™
S-3-600-8 |
203 mm | 2.41 mm | 0.006 mm | 0.0040 mm |
181.00 €
Pack of 5 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
12860026
|
Norell™
S-5-500-7 |
178 mm | 4.2 mm | 0.013 mm | 0.0050 mm |
151.00 €
Pack of 5 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
12830036
|
Norell™
S-5-200-8 |
203 mm | 4.2 mm | 0.035 mm | 0.0100 mm |
57.10 €
Pack of 5 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||
12890026
|
Norell™
S-5-400-8 |
203 mm | 4.2 mm | 0.019 mm | 0.0070 mm |
123.00 €
Pack of 5 |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
12870026
|
Norell™
S-5-500-8 |
203 mm | 4.2 mm | 0.013 mm | 0.0050 mm |
181.00 €
Pack of 5 |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||
Description
- Specifically designed for high-resolution NMR, tubes in the Select Series™ are manufactured from ASTM Type 1 Class A borosilicate glass
- Whether spinning or non-spinning, doing structure determination of small molecules or determining the 3D structure of a membrane protein, the Norell™ Select Series™ of NMR tubes offers unsurpassed performance, reproducibility and reliability compared to other brands
- You should see an immediate improvement in resolution when calculating i-coupling constants or acquiring data close to your spectrophotometer noise level, resulting in improved peak symmetry and shimmering times
- In turn this leads to shorter analysis times and improved sample throughput
- There are also no worries with freeze/thaw degassing or high temperature studies, these tubes can tolerate thermal shock up to 200°C
- All tubes have polished ends making them less susceptible to breaking
- Colored end caps are included
Specifications
Borosilicate Glass | |
NMR Sample Tube | |
5 Pack | |
Caps |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Norell™ Ultra Precision Borosilicate Glass NMR Sample Tubes