missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ NovaClean™ Lab and Glass Cleaner
BIOPHARMAGREENER_CHOICE

Ready to use, low-ionic detergent can be applied directly to a variety of cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains.

328.00 € - 384.00 €

Specifications

Product Type Filtered Low Ionic Surfactant (0.1 μ)
Color-coded No
pH 7.1
Product Line NovaClean™
Scent Unscented
View More Specs

Products 2
Product Code Brand Volume (English) Price Quantity & Availability  
Product Code Brand Volume (English) Price Quantity & Availability  
11966855
BIOPHARMA
View Documents
Micronova™
NC2-Q
1 qt. N/A N/A N/A
11962475
BIOPHARMAGREENER_CHOICE
View Documents
Micronova™
NC2-G
1 gal.
328.00 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ready to use, low-ionic detergent can be applied directly to a variety of cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains.

Lab and glass cleaner is a low-ionic detergent, designed for use on cleanroom work-stations, laminar flow benches, acrylic and painted surfaces.

  • Ready to use, cleanroom surfactant
  • Filtered to 0.1 microns
  • Low mobile ions
  • Low non-volatile residue levels
  • Can be applied directly to a range of surfaces
  • Works well with glass, stainless steel, epoxy, acrylic, and vinyl curtains
  • Ideal for conductive flooring
  • Available in a 1 quart spray bottle and a 1 gallon refill bottle
  • Biodegradable
  • Free-rinsing
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months

SDS
Documents

Documents

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Micronova™ NovaClean™ Lab and Glass Cleaner

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado