Learn More
Micronova™ NovaClean™ Lab and Glass Cleaner
BIOPHARMAGREENER_CHOICE
Ready to use, low-ionic detergent can be applied directly to a variety of cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains.
Brand:  Micronova™ NC2-G
284.48 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
 View more versions of this product
Product Code. 11962475
  • 328.00 € / Pack of 4
Explore more special offers
Description
Description
Ready to use, low-ionic detergent can be applied directly to a variety of cleanroom work surfaces including glass, stainless steel, epoxy acrylic, and vinyl curtains.
Lab and glass cleaner is a low-ionic detergent, designed for use on cleanroom work-stations, laminar flow benches, acrylic and painted surfaces.
  • Ready to use, cleanroom surfactant
  • Filtered to 0.1 microns
  • Low mobile ions
  • Low non-volatile residue levels
  • Can be applied directly to a range of surfaces
  • Works well with glass, stainless steel, epoxy, acrylic, and vinyl curtains
  • Ideal for conductive flooring
  • Available in a 1 quart spray bottle and a 1 gallon refill bottle
  • Biodegradable
  • Free-rinsing
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Micronova™ NovaClean™ Lab and Glass Cleaner > 1 gallon