Learn More
Emma Safety Footwear Novara Safety Shoes
Classic Brogue design with little more ankle support thanks to its extended shank. Virtually impenetrable nitrile rubber soles combined with the shoe's sophisticated looks, allow the wearer to move easily between the office and warehouse.
Brand: Emma Safety Footwear MM13109044
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather / Nubuck construction
- Chemical resistant / Liquid repellant
- Nitrile Rubber Heat Resistant Outsole
- ESD certified
- Chrome-free lining
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Brown | |
Hydro-Tec™ Sanitized Silver & Wellness leather | |
44 |
Laces | |
Novara | |
Leather, Nubuck | |
Steel |