Learn More
Emma Safety Footwear Novara Safety Shoes
Classic Brogue design with little more ankle support thanks to its extended shank. Virtually impenetrable nitrile rubber soles combined with the shoe's sophisticated looks, allow the wearer to move easily between the office and warehouse.
Brand:  Emma Safety Footwear MM13109047
 View more versions of this product
Product Code. 15978211
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather / Nubuck construction
  • Chemical resistant / Liquid repellant
  • Nitrile Rubber Heat Resistant Outsole
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Brown
Hydro-Tec™ Sanitized Silver & Wellness leather
47
Laces
Novara
Leather, Nubuck
Steel