Learn More
Invitrogen™ Novex™ Tricine Mini Protein Gels, 10 to 20%, 1.0 mm
Invitrogen Novex Tricine Gels provide separation of low molecular weight proteins and peptides. In this system tricine replaces glycine in the running buffer, resulting in more efficient stacking and destacking for low molecular weight proteins and higher resolution of smaller peptides.
Brand: Invitrogen™ EC66252BOX
202.92 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: Catalog numbers for gels refer to a single box. One box contains 10 gels.
Description
Invitrogen Novex Tricine Gels provide separation of low molecular weight proteins and peptides. The Tricine system is a modification of the tris-glycine discontinuous buffer system developed by Schaegger and von Jagow (Schaegger and von Jagow, 1987) specifically for resolving peptides and low molecular weight proteins. In this system the tricine replaces the glycine in the running buffer, resulting in more efficient stacking and destacking for low molecular weight proteins and higher resolution of smaller peptides.
Features of Novex Tricine protein gels:
• Increased resolution of proteins with molecular weights as low as 2 kDa
• Improved compatibility with direct sequencing of proteins after transferring to PVDF
• Minimized protein modification due to the lower pH of the tricine buffering system
Formulation
Invitrogen Tricine gels are made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, and highly purified water. Our Tricine gels have a 4% stacking gel and do not contain SDS. The Tricine system requires SDS in sample and running buffers for best results.
Choose the right Tricine gel for your protein separation
Invitrogen Tricine gels come in three polyacrylamide concentrations of 10%, 16%, and a gradient of 10–20%. Select from our many well formats, including 10-, 12-, and 15-well. Tricine gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, we recommend Tricine SDS Sample Buffer (LC1676) and optimal separation use Tricine SDS Running Buffer (LC1675).
For transfer of proteins to a membrane, we recommend using the Novex Tris-Glycine Transfer Buffer (LC3675) if performing a traditional wet transfer using the XCell II Blot Module (EI9051) or the Mini Blot Module (B1000). Rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).
Order Info
Shipping Condition: Wet Ice

Specifications
Mini Gel Tank, XCell SureLock Mini-Cell | |
Mini | |
Tricine | |
Up to 20 μL | |
Denaturing | |
Molecular Weight | |
10 Gels/Box | |
Store at 2°C to 8°C. Do not freeze. | |
8 cm |
10 to 20% | |
1.0 mm | |
Denaturing | |
2.5 to 250 kDa | |
12-well | |
Novex™ | |
Wet Ice | |
8 cm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
18103141 | Certificate of Analysis | 06/11/2018 | EC66252BOX,EC6625BOX |
12121943 | Certificate of Analysis | 19/07/2017 | EC66252BOX |
12112943 | Certificate of Analysis | 19/07/2017 | EC66252BOX |
12110746 | Certificate of Analysis | 19/07/2017 | EC66252BOX |
13010243 | Certificate of Analysis | 19/07/2017 | EC66252BOX |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.