missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 10X Bolt™ Sample Reducing Agent

Optimized for use with Bolt™ Bis-Tris Plus gels, and available in a variety of formats.

Brand:  Invitrogen™ B0009

51.65 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 13296499

  • 75.00 € / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 500mM dithiothreitol (DTT)
This item is not returnable. View return policy

Description

Description

  • Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of small- to medium-sized proteins under denaturing conditions
  • Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications
  • Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial
  • Also use Bolt™ gels to obtain optimal results for day-to-day protein separation needs
  • Gels come in four gel types and multiple well formats
  • A Mini Gel Tank is required for use of all Bolt™ gels
  • Run times may vary depending upon the type of running buffer used
  • Store at 2°C to 8°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sample Reducing Agent (10X) contains 500 mM dithiothreitol (DTT).

Store at 2°C to 8°C.
Bolt Mini Gel Tank
Liquid
Bolt™ Bis-Tris Plus Gels
10 mL
10X, 500 mM
Sample Reducing Agent
500 mM dithiothreitol (DTT)
Bolt™
Reducing Agent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3110542 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberB0009,B0009_LITL18 PROMO
Lot Number3033876 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberB0009,B0009_LITL18 PROMO
Lot Number2459664 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberB0009,B0009_LITL18 PROMO
Lot Number2495541 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberB0009,B0009_LITL18 PROMO
Lot Number2501135 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberB0009,B0009_LITL18 PROMO
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ 10X Bolt™ Sample Reducing Agent > 10 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.