missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ TE42 and TE62 Tank Blotting Units
Perform high-intensity protein transfers in one hour
Brand: Hoefer™ TE42
This item is not returnable.
View return policy
Description
- Over 1.5 m of platinum wire per electrode provides uniform field strength for efficient transfers
- Hinged, color-coded cassettes enable simple loading and easy orientation of gel sandwich
- Buffer volumes of 4 to 5 L required, depending on number of cassettes processed
- Model TE62 offers a built-in heat exchanger in the base of the unit
- Model TE42 can be used with optional heat exchanger (Mfr. No. TE47)
Certifications
CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1
Specifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE | |
Standard Transfer Tank | |
Lower chamber, safety lid with high voltage Leads, 2 electrode panels, 2 gel cassettes, four 0.13 in. thick Foam sponges, two 0.25 in. thick Foam sponges, 25 sheets blotting Paper, cassette hook | |
45°C | |
4000 to 5000 mL | |
12 in. | |
11.02 in. | |
5.11 in. | |
100 V | |
2 A |
Up to 2, 15 x 21 cm Gel (with cooling), up to 4, 15 x 21 cm Gel (without cooling) | |
Up to 80% | |
113°F | |
135.25 to 169.07 oz. | |
12 x 11.02 x 5.11 in. (30.5 x 28 x 13 cm) | |
30.5 cm | |
28 cm | |
13 cm | |
200 w |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
HOE001 Hoefer Catalogue 2009 10
File size: 9.7 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hoefer™ TE42 and TE62 Tank Blotting Units > Four 0.12 in. thick foam sponges
Spot an opportunity for improvement?Share a Content Correction