missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cell Extraction Buffer

Suitable for use in ELISA and Western blotting

Brand:  Invitrogen™ FNN0011

226.10 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10009222

  • 244.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Build Your Own Immunoassay, Cell Analysis, Cell Lysis, Immunoassays, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C).
Western Blot, Sample preparation for ELISA
Dry Ice
Cell Extraction Buffer
100 ml
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Cell Extraction Buffer > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.