Learn More
Invitrogen™ Novex™ ECL Chemiluminescent Substrate Reagent Kit
Chemiluminescent substrate used for chemiluminescence-based immunodetection of horse radish peroxidase (HRP) on western or dot blot membranes
Brand: Invitrogen™ WP20005
272.65 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
It is a two-part reagent consisting of Reagent A (luminol) and Reagent B (an enhancer) used in equal volumes to attain the most intense light emission.
Order Info
Approved for shipment at Room Temperature or on Wet Ice

Specifications
2 x 125 mL bottles | |
Chemical Substrate | |
Comes with two bottles. Reagent A (125 ml) and Reagent B (125 ml). Avoid skin contact. Store at 4°C. |
|
Liquid | |
Luminol Substrate |
Approved for shipment at Room Temperature or on Wet Ice | |
HRP (Horseradish Peroxidase) Substrate | |
Chemiluminescence | |
Novex™ |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3005658 | Certificate of Analysis | 31/01/2025 | WP20005 |
2459666 | Certificate of Analysis | 29/01/2025 | WP20005 |
2563468 | Certificate of Analysis | 29/01/2025 | WP20005 |
2631801 | Certificate of Analysis | 29/01/2025 | WP20005 |
2822763 | Certificate of Analysis | 28/01/2025 | WP20005 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.