missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gel-Dry™ Drying Solution

Modulates the rate of water release to ensure crack-free gels

Brand:  Invitrogen™ LC4025


 View more versions of this product

Product Code. 11599066

  • 46.67 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Gel-Dry™ Drying Solution is one component of the DryEase™ Mini-Gel Drying System which is a simple, easy-to-use kit for drying gels evenly using passive evaporation
  • The kit allows gels to be assembled on a base between two open frames, so that both outer surfaces of the gels are exposed to air
  • Consequently, drying is even and cracking is minimized
  • The proprietary Gel-Dry™ Drying Solution modulates the rate of water release to ensure crack-free gels
  • With the DryEase™ System, gels are dried overnight with uniform tension on all points of the gel to eliminate cracking

Applications

  • Gels dried with the DryEase™ System are suitable for fluorography, densitometry, autoradiography, or permanent gel storage

Order Info

Shipping condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Drying Solutions
DryEase™ Mini-Gel Drying System
Novex
500 mL
500 mL
Store at room temperature.
Mini
Drying Solution
Room Temperature
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Gel-Dry Drying Solution

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.

Supplemental information
  • MIXTURE LIST-Contains : Ethanol, Methanol, Propan-2-ol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.