missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ HRP Chromogenic Substrate (TMB)

Ready to use single component reagent used for sensitive detection of a horse radish peroxidase (HRP) on western blot and dot blot membranes

Brand:  Invitrogen™ WP20004

Product Code. 10388413

  • 214.00 € / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Consisting of a ready-to-use solution of TMB, the Novex™ HRP Chromogenic Substrate forms a blue precipatate up reaction with horse radish peroxidase (HRP).

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 x 250 ml bottle
Chemical Substrate
Blue
Colorimetric
Novex™
Approved for shipment at Room Temperature or on Wet Ice
HRP (Horseradish Peroxidase) Substrate
Single bottle labelled: Novex™ HRP Chromogenic Substrate (TMB). Avoid exposure to light. Store at 4°C.
Liquid
TMB
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Novex™ HRP Chromogenic Substrate (TMB) > 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.