Learn More
Invitrogen™ NuPAGE™ Transfer Buffer (20X)
Use to transfer proteins from NuPAGE Novex gels to membranes for Western blotting
48.27 € - 280.00 €
Specifications
Content And Storage | Store at room temperature. Guaranteed stable for 6 months unless otherwise specified in the product literature. |
---|---|
Buffer | Transfer Buffers |
Gel Type | NuPAGE |
Product Type | Transfer Buffer |
Gel Compatibility | NuPAGE™ Gels |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11539166
|
Invitrogen™
NP0006 |
125mL |
48.27 €
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10564613
|
Invitrogen™
NP00061 |
1L |
280.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NuPAGE Transfer Buffer can be used with the Novex Semi-Dry Blotter for semi-dry transfer or with the XCell II™ Blot Module for wet (tank) transfer. NuPAGE Antioxidant may be used with NuPAGE Transfer Buffer to enhance transfer of reduced proteins to membranes.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature. Guaranteed stable for 6 months unless otherwise specified in the product literature. | |
NuPAGE | |
NuPAGE™ Gels | |
Room Temperature |
Transfer Buffers | |
Transfer Buffer | |
NuPAGE™ |
Safety and Handling
missing translation for 'shelfLife' : 6 Months
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.