Learn More
Invitrogen™ Novex™ IEF Protein Gels, pH 3-7, 1.0 mm
Novex™ pH 3-7 IEF Gels are used for pI determination and are excellent for native applications using soluble proteins. Isoelectric focusing (IEF) is an electrophoresis technique that separates proteins based on their isoelectric point (pI).
Brand: Invitrogen™ EC6645BOX
228.00 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Novex™ pH 3-7 IEF Gels are used for pI determination and are excellent for native applications using soluble proteins. Isoelectric focusing (IEF) is an electrophoresis technique that separates proteins based on their isoelectric point (pI). The pI is the pH at which a protein has no net charge and does not move in an electric field. Vertical IEF gels effectively create a pH gradient so proteins separate according to their unique pI.
IEG gels can be used to readily detect minor changes in a protein due to deamination, phosphorylation, or glycosylation, and can resolve different proteins of similar size that cannot be resolved on standard SDS-PAGE gels. Pre-focusing is not required. Total run time is approximately 2.5 hours.
Formulation
Novex™ IEF gels are 5% polyacrylamide and consist of high-purity acrylamide, bisacrylamide, TEMED, APS, ultrapure water, and 2% ampholytes. They do not contain denaturing reagents.
Recommended buffers
IEF is a sensitive technique which is affected by many factors. Our optimized, premixed IEF buffers reduce variability and help you obtain consistent results.

Specifications
Mini Gel Tank, XCell SureLock Mini-Cell | |
Mini | |
IEF | |
25 μL | |
10-well | |
Novex™ | |
Wet Ice | |
8 cm |
5% | |
1.0 mm | |
3.5 to 6.8 | |
Isoelectric Focusing | |
pI (Charge) | |
5 Gels/Box | |
Store at 2°C to 8°C. Do not freeze. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
18020141 | Certificate of Analysis | 26/07/2018 | EC6645BOX,EC66452BOX |
12121435 | Certificate of Analysis | 19/07/2017 | EC6645BOX |
12112633 | Certificate of Analysis | 19/07/2017 | EC6645BOX |
13053151 | Certificate of Analysis | 14/04/2017 | EC6645BOX |
13071751 | Certificate of Analysis | 13/04/2017 | EC6645BOX |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.