missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine SDS Running Buffer (10X)

Formulated for separation of proteins, in their denatured state, on Novex Tris-Glycine gels

Brand:  Invitrogen™ LC2675

34.16 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 11559066

  • 56.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Novex Tris-Glycine SDS Running Buffer (10X), 500mL

Requires : The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.
This item is not returnable. View return policy

Description

Description

Tris-Glycine gels provide reproducible separation of a wide range of proteins into well-resolved bands, depending upon the gel percentage used.

  • Pre-mixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results
  • Made with high-purity reagents and are strictly quality controlled

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Running Buffer
Liquid
500 mL
10X
Novex
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3005618 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberLC2675,LC2675_LITL18 PROMO
Lot Number3071212 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberLC2675,LC2675_LITL18 PROMO
Lot Number2480833 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberLC2675,LC2675_LITL18 PROMO
Lot Number2480834 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberLC2675,LC2675_LITL18 PROMO
Lot Number2493864 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberLC2675,LC2675_LITL18 PROMO
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Novex™ Tris-Glycine SDS Running Buffer (10X) > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.