Learn More
Invitrogen™ Novex™ Tris-Glycine SDS Running Buffer (10X)
Formulated for separation of proteins, in their denatured state, on Novex Tris-Glycine gels
54.25 € - 293.00 €
Specifications
Chemical Name or Material | Running Buffer |
---|---|
Concentration | 10X |
Product Line | Novex |
Includes: Novex Tris-Glycine SDS Running Buffer (10X), 500mL
Requires: The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11559066
|
Invitrogen™
LC2675 |
500 mL |
54.25 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10040173
|
Invitrogen™
LC26754 |
4 x 1 L |
266.00 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11569066
|
Invitrogen™
LC26755 |
5 L |
293.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tris-Glycine gels provide reproducible separation of a wide range of proteins into well-resolved bands, depending upon the gel percentage used.
- Pre-mixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results
- Made with high-purity reagents and are strictly quality controlled
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Running Buffer | |
10X | |
Novex |
Safety and Handling
Recommended Storage : Contains: Novex™ Tris-Glycine SDS Running Buffer (10X), 500 ml
Storage: +4°C to 25°C
Guaranteed stable for 6 months unless otherwise specified in the product literature.
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.