missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals NP-40 lysis buffer

Brand:  Thermo Scientific Chemicals J60766.AK

 View more versions of this product

Product Code. 15493479

  • 69.14 € / 250mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.

Solubility
It is soluble in water.

Notes
This buffer contains: 50mM Tris-HCl (pH 7.4), 150mM NaCl, 1%NP-40 and 5mM EDTA. To use, incubate cells in 5 volumes of buffer on ice for 10-30 minutes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorless
Liquid
It is soluble in water.
Keep cold
250 mL
Lysis Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only