Learn More
Thermo Scientific Chemicals NP-40 lysis buffer
69.14 € - 97.79 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15403489
|
Thermo Scientific Chemicals
J60766.AP |
500 mL |
97.79 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15493479
|
Thermo Scientific Chemicals
J60766.AK |
250 mL |
69.14 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNP40 Cell Lysis Buffer is suitable for the preparation of cell extracts to be analyzed by Antibody Bead Immunoassay (Luminex), ELISA, and Western blotting. It can also be used as a wash buffer for immunoprecipitation reactions.
Solubility
It is soluble in water.
Notes
This buffer contains: 50mM Tris-HCl (pH 7.4), 150mM NaCl, 1%NP-40 and 5mM EDTA. To use, incubate cells in 5 volumes of buffer on ice for 10-30 minutes.
Specifications
Colorless | |
Keep cold | |