Learn More
Cytiva nProtein A Sepharose™ 4 Fast Flow Affinity Media
Used for purification of monoclonal and polyclonal antibodies. Cytiva Lifescience™ nProtein A Sepharose™ 4 Fast Flow Affinity Media contains native protein A coupled to Sepharose 4 Fast Flow.
Brand:  Cytiva 17-5280-01
 View more versions of this product
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11359931
  • 756.00 € / 5mL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Low leakage of protein A with nearly twice total IgG binding capacity of Protein A Sepharose CL-4B
  • Used in large-scale FDA-approved processes
  • Manufactured without using animal-derived components
  • Rigidity and high degree of substitution of Sepharose 4 Fast Flow matrix facilitates speedy process of large volumes dilute cell culture fluid
  • High chemical, thermal and mechanical stability
  • Withstands high concentrations of hydrogen bond disrupting agents
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 mL
For PurifiCatIon of Monoclonal and Polyclonal Antibodies
>35 mg Human IgG/mL
150 to 250 cm/Hoμr
9.84 in.
100 kPa
3 to 9
Native Protein A (S. aureus)
No significant change in chromatographic performance after 1 week storage using 8 M Urea, 6 M Gua-HCI, 2% Benzyl Alcohol or 20% Ethanol
2°C to 8°C
nProtein A Sepharose 4 Fast Flow Media
90 μm
25 cm
4% Cross-Linked Agarose
20% Ethanol