missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva nProtein A Sepharose™ 4 Fast Flow Affinity Media
Used for purification of monoclonal and polyclonal antibodies. Cytiva Lifescience™ nProtein A Sepharose™ 4 Fast Flow Affinity Media contains native protein A coupled to Sepharose 4 Fast Flow.
756.00 € - 2174.96 €
Specifications
Stability | No significant change in chromatographic performance after 1 week storage using 8 M Urea, 6 M Gua-HCI, 2% Benzyl Alcohol or 20% Ethanol |
---|---|
Storage Requirements | 2°C to 8°C |
Binding Capacity | >35 mg Human IgG/mL |
Product Type | nProtein A Sepharose 4 Fast Flow Media |
Flow Rate | 150 to 250 cm/Hoμr |
Product Code | Brand | For Use With (Application) | pH Range | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | pH Range | Quantity | Price | Quantity & Availability | |||
11359931
|
Cytiva
17-5280-01 |
For PurifiCatIon of Monoclonal and Polyclonal Antibodies | 3 to 9 | 5 mL |
756.00 €
5mL |
In Stock
Log in to see stock. |
|||
11379931
|
Cytiva
17-5280-04 |
For The PurifiCatIon of Monoclonal and Polyclonal Antibodies at Both Laboratory and Process Scale | 2 to 10 | 25 mL | N/A | ||||
Description
- Low leakage of protein A with nearly twice total IgG binding capacity of Protein A Sepharose CL-4B
- Used in large-scale FDA-approved processes
- Manufactured without using animal-derived components
- Rigidity and high degree of substitution of Sepharose 4 Fast Flow matrix facilitates speedy process of large volumes dilute cell culture fluid
- High chemical, thermal and mechanical stability
- Withstands high concentrations of hydrogen bond disrupting agents
Specifications
No significant change in chromatographic performance after 1 week storage using 8 M Urea, 6 M Gua-HCI, 2% Benzyl Alcohol or 20% Ethanol | |
>35 mg Human IgG/mL | |
150 to 250 cm/Hoμr | |
9.84 in. | |
100 kPa | |
20% Ethanol |
2°C to 8°C | |
nProtein A Sepharose 4 Fast Flow Media | |
90 μm | |
25 cm | |
4% Cross-Linked Agarose | |
Native Protein A (S. aureus) |