missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSE2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564243
This item is not returnable.
View return policy
Description
Immunogen sequence: QNNEKFVQFK QQLKELKKQC GLQADREADG TEGVDEDIIV TQS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.
This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients.
Specifications
NSE2 | |
Polyclonal | |
Unconjugated | |
Nsmce2 | |
1110014D18Rik; AI661537; C8orf36; E3 SUMO-protein ligase NSE2; E3 SUMO-protein transferase NSE2; FLJ32440; hMMS21; methyl methanesulfonate sensitivity gene 21; MMS21; MMS21 homolog; non-SMC element 2 homolog; non-SMC element 2 homolog (MMS21, S. cerevisiae); non-SMC element 2, MMS21 homolog; non-SMC element 2, MMS21 homolog (S. cerevisiae); non-structural maintenance of chromosomes element 2 homolog; NSE2; NSE2 (MMS21) homolog, SMC5-SMC6 complex SUMO ligase; NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase; NSMCE2; RGD1305156; Unknown (protein for MGC:133996); zinc finger, MIZ-type containing 7; ZMIZ7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286053 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96MF7 | |
Nsmce2 | |
Recombinant protein corresponding to Human NSMCE2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NSE2 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Research Use Only
Spot an opportunity for improvement?Share a Content Correction