missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ PC EF Kit Buffer Solutions
Additional supply of buffer for use with NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100) for the purification of endotoxin-free plasmid DNA. Buffer solutions include: S1/S2/S3-EF, N2/N3/N4/N5-EF, TE-EF, and H2O-EF.
Brand: Macherey-Nagel™ 740795.1
This item is not returnable.
View return policy
Description
NucleBond™ PC EF: The classic anion exchanger for purification of endotoxin-free plasmid DNA
- Proven anion-exchange technology – transfection-grade plasmid DNA
- Plasmid DNA with less than 0.1 EU/μg for transfection of highly sensitive cells
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Fast and easy clarification of lysates by Folded Filters – no centrifugation required, no shearing forces
- NucleoBond™ Finalizers available to avoid time consuming centrifugation for plasmid precipitation
- Separate binding columns of NucleoBond™ PC kits available as NucleoBond™ AX columns
Specifications
Colorless | |
NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100) | |
6.0 to 6.5 | |
64-17-5d | |
Room Temperature (18°C to 25°C) | |
Purification of endotoxin-free plasmid DNA |
Ethanol, diluted < 20%; Potassium chloride; Water | |
Ethanol, diluted < 20%; Potassium chloride; Water | |
1000 mL | |
Alcoholic | |
Liquid | |
Wash Buffer N4-EF |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Buffer N4-EF (1000 mL)
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction