Learn More
Macherey-Nagel™ NucleoBond™ PC EF Kit Buffer Solutions
Additional supply of buffer for use with NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100) for the purification of endotoxin-free plasmid DNA. Buffer solutions include: S1/S2/S3-EF, N2/N3/N4/N5-EF, TE-EF, and H2O-EF.
53.00 € - 143.00 €
Specifications
Color | Colorless |
---|---|
For Use With (Equipment) | NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100) |
CAS | 64-17-5d |
Content And Storage | Room Temperature (18°C to 25°C) |
Form | Liquid |
Product Code | Brand | Buffer | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Buffer | Quantity | Price | Quantity & Availability | ||||
15869107
|
Macherey-Nagel™
740790.1 |
1000 mL |
75.90 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15879107
|
Macherey-Nagel™
740791.1 |
1000 mL |
75.90 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15889107
|
Macherey-Nagel™
740792.1 |
1000 mL |
143.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15899107
|
Macherey-Nagel™
740793.1 |
1000 mL |
104.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15809117
|
Macherey-Nagel™
740794.1 |
1000 mL |
104.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15819117
|
Macherey-Nagel™
740795.1 |
1000 mL |
104.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15829117
|
Macherey-Nagel™
740796.1 |
600 mL |
104.00 €
600mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15839117
|
Macherey-Nagel™
740797.1 |
1000 mL |
88.75 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15849117
|
Macherey-Nagel™
740798.1 |
1000 mL |
53.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NucleBond™ PC EF: The classic anion exchanger for purification of endotoxin-free plasmid DNA
- Proven anion-exchange technology – transfection-grade plasmid DNA
- Plasmid DNA with less than 0.1 EU/μg for transfection of highly sensitive cells
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Fast and easy clarification of lysates by Folded Filters – no centrifugation required, no shearing forces
- NucleoBond™ Finalizers available to avoid time consuming centrifugation for plasmid precipitation
- Separate binding columns of NucleoBond™ PC kits available as NucleoBond™ AX columns
Specifications
Colorless | |
64-17-5d | |
Liquid |
NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100) | |
Room Temperature (18°C to 25°C) | |
Purification of endotoxin-free plasmid DNA |
Safety and Handling
missing translation for 'shelfLife' : At least 1 year
Your input is important to us. Please complete this form to provide feedback related to the content on this product.