Learn More
Macherey-Nagel™ NucleoBond™ PC EF Kit Buffer Solutions
Additional supply of buffer for use with NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100) for the purification of endotoxin-free plasmid DNA. Buffer solutions include: S1/S2/S3-EF, N2/N3/N4/N5-EF, TE-EF, and H2O-EF.
Brand:  Macherey-Nagel™ 740792.1
 View more versions of this product
Product Code. 15889107
  • 143.00 € / 1 L
Explore more special offers
Description
Description
NucleBond™ PC EF: The classic anion exchanger for purification of endotoxin-free plasmid DNA
  • Proven anion-exchange technology – transfection-grade plasmid DNA
  • Plasmid DNA with less than 0.1 EU/μg for transfection of highly sensitive cells
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Fast and easy clarification of lysates by Folded Filters – no centrifugation required, no shearing forces
  • NucleoBond™ Finalizers available to avoid time consuming centrifugation for plasmid precipitation
  • Separate binding columns of NucleoBond™ PC kits available as NucleoBond™ AX columns
Specifications
Specifications
Colorless
NucleoBond™ PC EF kits (PC 500 EF, PC 2000 EF, PC 10000 EF, PC Prep 100)
5.2 to 5.7
64-17-5d
Room Temperature (18°C to 25°C)
Purification of endotoxin-free plasmid DNA
Acetic acid; Potassium; Sodium + Water
Acetic acid; Potassium; Sodium + Water
1000 mL
Acetic
Liquid
Acetate buffer solution