missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ RNA Soil Mini

Easy handling and superior speed in RNA purification from soil samples for qRT-PCR analysis, as well as parallel preparation of DNA. Complete kit uses anion exchange chromatography and gravity flow columns combined with NucleoSpin Bead Tubes Type A (included).

86.25 € - 940.00 €

Specifications

Format Gravity Flow Column
Isolation Technology Anion Exchange Chromatography, Gravity Flow Columns, NucleoSpin Bead Tubes
Elution Volume Elution volume: 50-100 μL
Sample Type Soil
Efficiency Binding capacity: 30 μg
View More Specs

Products 3
Product Code Brand No. of Reactions Product Type Price Quantity & Availability  
Product Code Brand No. of Reactions Product Type Price Quantity & Availability  
15858096
View Documents
Macherey-Nagel™
740142.10
10 RNA Kit
203.00 €
10 reactions
Please to purchase this item. Need a web account? Register with us today!
15868096
missing translation for 'viewDocuments'
Macherey-Nagel™
740142.50
50 RNA Kit
940.00 €
50 reactions
Please to purchase this item. Need a web account? Register with us today!
15937282
missing translation for 'viewDocuments'
Macherey-Nagel™
740142.2
2 RNA Kit + DNA Set
86.25 €
2 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NucleoBond® RNA Soil Mini kit enables efficient isolation of high quality RNA and DNA from any kind of soil or sediment sample. Most soils contain relatively low amounts of RNA, compared to DNA. Therefore, the kit is designed with proven NucleoBond™ technology to enable high sample input and and pure nucleic acids.

  • Anion exchange technology to optimize RNA yield and purity—suitable for qRT-PCR analysis
  • Combination of mechanical homogenization and chemical lysis allows processing of large samples
  • Parallel preparation of RNA and DNA in one hour*
  • Sample disruption is achieved with NucleoSpin™ Bead Tubes Type A, optimized Lysis Buffer, and Phenol:Chloroform:Isoamylalcohol
  • Optional Buffer OPT enables RNA and DNA purification from mineralic and organic soil samples

* DNA isolation requires the DNA Set for NucleoBond™ RNA Soil Mini, sold separately (man. part no. 740143.10 or 740143.50) or in a combo kit

Specifications

Specifications

Gravity Flow Column
Elution volume: 50-100 μL
Binding capacity: 30 μg
RNA/DNA isolation from soil for qRT-PCR
A260/A280: 1.5 - 2.0
RNA integrity number (RIN): > 7.0
Anion Exchange Chromatography, Gravity Flow Columns, NucleoSpin Bead Tubes
Soil
Centrifuge for microcentrifuge tubes
NucleoBond Soil Mini Columns, NucleoSpin Bead Tubes Type A, NucleoSpin RNA columns, Buffers
< 0.25 - 0.5 g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • NucleoBond RNA Soil Mini (10)

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Supplemental information
  • MIXTURE LIST-Contains : EDTA, tris(hydroxymethyl)aminomethane, sodium chloride, dodecyl sulphate sodiumsalt, potassium chlo

SDS
Documents

Documents

Certificates