Learn More
Macherey-Nagel™ Nucleodur™ C18 Isis HPLC Column
Brand:  Macherey-Nagel™ 760404.46
 View more versions of this product
Product Code. 12428437
  • 979.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Polymeric octadecyl
Polymeric octadecyl
1 mL/min
250 mm
Acetonitrile/water
3 μm
110 Å
1 Ea.
340 m²/g
L1
Silica
Steric Selectivity
Reversed Phase
EC Column
Analytical Column
4.6 mm
For Steroids, (o,p,m-) Substituted Aromatics, Fat-Soluble Vitamins Application
8700 psi (600 bar)
Spherical
1 to 10
0.9 mL/g
Acetonitrile/water
60°C
C18 Isis
20%
Yes
Stainless Steel