Learn More
Macherey-Nagel™ Nucleodur™ C18 Isis HPLC Column
625.00 € - 925.00 €
Specifications
Column Format | Analytical Column |
---|---|
Particle Shape | Spherical |
Pore Size | 110 Å |
Quantity | 1 Ea. |
Solvents | Acetonitrile/water |
Product Code | Brand | Diameter (Metric) | Particle Size | Length (Metric) | Carbon Load | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Particle Size | Length (Metric) | Carbon Load | Price | Quantity & Availability | ||
12757830
|
Macherey-Nagel™
760409.20 |
2 mm | 1.8 μm | 150 mm | 20% |
770.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12418447
|
Macherey-Nagel™
760407.20 |
2 mm | 1.8 μm | 100 mm | 20% |
770.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12418427
|
Macherey-Nagel™
760402.20 |
2 mm | 3 μm | 125 mm | 20% |
770.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12458427
|
Macherey-Nagel™
760403.20 |
2 mm | 3 μm | 150 mm | 20% |
770.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12498427
|
Macherey-Nagel™
760404.20 |
2 mm | 3 μm | 250 mm | 20% |
890.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12478437
|
Macherey-Nagel™
760406.20 |
2 mm | 1.8 μm | 30 mm | 20% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12438437
|
Macherey-Nagel™
760405.20 |
2 mm | 1.8 μm | 50 mm | 20% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12468417
|
Macherey-Nagel™
760400.20 |
2 mm | 3 μm | 50 mm | 20% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12737830
|
Macherey-Nagel™
760396.20 |
2 mm | 1.8 μm | 75 mm | 20% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12428427
|
Macherey-Nagel™
760402.30 |
3 mm | 3 μm | 125 mm | 20% |
780.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12468427
|
Macherey-Nagel™
760403.30 |
3 mm | 3 μm | 150 mm | 20% |
780.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12408437
|
Macherey-Nagel™
760404.30 |
3 mm | 3 μm | 250 mm | 20% |
885.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12488437
|
Macherey-Nagel™
760406.30 |
3 mm | 1.8 μm | 30 mm | 20% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12448437
|
Macherey-Nagel™
760405.30 |
3 mm | 1.8 μm | 50 mm | 20% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12478417
|
Macherey-Nagel™
760400.30 |
3 mm | 3 μm | 50 mm | 20% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12408427
|
Macherey-Nagel™
760401.46 |
4.6 mm | 3 μm | 100 mm | 20% |
840.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12448427
|
Macherey-Nagel™
760402.46 |
4.6 mm | 3 μm | 125 mm | 20% |
840.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12488427
|
Macherey-Nagel™
760403.46 |
4.6 mm | 3 μm | 150 mm | 20% |
840.00 €
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
||
12408447
|
Macherey-Nagel™
760406.46 |
4.6 mm | 1.8 μm | 30 mm | 20% |
735.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12468437
|
Macherey-Nagel™
760405.46 |
4.6 mm | 1.8 μm | 50 mm | 20% |
735.00 €
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||
12498417
|
Macherey-Nagel™
760400.46 |
4.6 mm | 3 μm | 50 mm | 20% |
735.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12428437
|
Macherey-Nagel™
760404.46 |
4.6 mm | 3 μm | 250 mm | 20% |
925.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12478507
|
Macherey-Nagel™
760755.30 |
3 mm | 1.8 μm | 50 mm | 11% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12747830
|
Macherey-Nagel™
760397.46 |
4.6 mm | 3 μm | 75 mm | 20% |
735.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12438427
|
Macherey-Nagel™
760402.40 |
4 mm | 3 μm | 125 mm | 20% |
780.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12488447
|
Macherey-Nagel™
760412.40 |
4 mm | 5 μm | 125 mm | 20% |
625.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12478427
|
Macherey-Nagel™
760403.40 |
4 mm | 3 μm | 150 mm | 20% |
780.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12418437
|
Macherey-Nagel™
760404.40 |
4 mm | 3 μm | 250 mm | 20% |
885.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12498437
|
Macherey-Nagel™
760406.40 |
4 mm | 1.8 μm | 30 mm | 20% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12458437
|
Macherey-Nagel™
760405.40 |
4 mm | 1.8 μm | 50 mm | 20% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
12488417
|
Macherey-Nagel™
760400.40 |
4 mm | 3 μm | 50 mm | 20% |
680.00 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
Specifica
Analytical Column | |
110 Å | |
Acetonitrile/water | |
60°C | |
Yes | |
EC Column |
Spherical | |
1 Ea. | |
340 m²/g | |
Silica | |
Stainless Steel |