Learn More
Macherey-Nagel™ Nucleodur™ Sphinx RP EC Analytical HPLC Column
Column Format: Analytical Column; Carbon Load: 15%; Pore Size: 110 Å; Column Phase: Reversed Phase
564.00 € - 795.00 €
Specifications
Column Chemistry | Silica |
---|---|
Column Format | Analytical Column |
Crosslinking | Octadecyl and phenyl |
For Use With | HPLC |
Functionality/Interaction Type | p-p Interactions |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
12498517
|
Macherey-Nagel™
760801.20 |
125 mm | 2 mm |
646.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
12438527
|
Macherey-Nagel™
760802.20 |
150 mm | 2 mm |
646.00 €
Each |
In Stock
Log in to see stock. |
||||
12458517
|
Macherey-Nagel™
760800.20 |
50 mm | 2 mm |
564.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
12408527
|
Macherey-Nagel™
760801.30 |
125 mm | 3 mm |
656.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
12478527
|
Macherey-Nagel™
760803.20 |
250 mm | 2 mm |
754.00 €
Each |
In Stock
Log in to see stock. |
||||
12448527
|
Macherey-Nagel™
760802.30 |
150 mm | 3 mm |
656.00 €
Each |
In Stock
Log in to see stock. |
||||
12488527
|
Macherey-Nagel™
760803.30 |
250 mm | 3 mm |
749.00 €
Each |
In Stock
Log in to see stock. |
||||
12468517
|
Macherey-Nagel™
760800.30 |
50 mm | 3 mm |
564.00 €
Each |
In Stock
Log in to see stock. |
||||
12428527
|
Macherey-Nagel™
760801.46 |
125 mm | 4.6 mm |
723.00 €
Each |
In Stock
Log in to see stock. |
||||
12468527
|
Macherey-Nagel™
760802.46 |
150 mm | 4.6 mm |
723.00 €
Each |
In Stock
Log in to see stock. |
||||
10524644
|
Macherey-Nagel™
760803.46 |
250 mm | 4.6 mm |
795.00 €
Each |
In Stock
Log in to see stock. |
||||
12458547
|
Macherey-Nagel™
760809.46 |
100 mm | 4.6 mm |
723.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
12488517
|
Macherey-Nagel™
760800.46 |
50 mm | 4.6 mm |
615.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
12665989
|
Macherey-Nagel™
760815.46 |
75 mm | 4.6 mm |
615.00 €
Each |
In Stock
Log in to see stock. |
||||
12418527
|
Macherey-Nagel™
760801.40 |
125 mm | 4 mm |
656.00 €
Each |
In Stock
Log in to see stock. |
||||
12458527
|
Macherey-Nagel™
760802.40 |
150 mm | 4 mm |
656.00 €
Each |
In Stock
Log in to see stock. |
||||
12498527
|
Macherey-Nagel™
760803.40 |
250 mm | 4 mm |
749.00 €
Each |
In Stock
Log in to see stock. |
||||
12478517
|
Macherey-Nagel™
760800.40 |
50 mm | 4 mm |
564.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
Specifications
Silica | |
Octadecyl and phenyl | |
p-p Interactions | |
8700 psi (600 bar) | |
5 μm | |
110 Å | |
Acetonitrile/water | |
60°C | |
Silica | |
Reversed Phase | |
Reversed Phase |
Analytical Column | |
HPLC | |
Yes | |
Acetonitrile/water | |
1 to 10 | |
1 Ea. | |
340 m²/g | |
C18 and Propylphenyl (Sphinx RP) | |
15% | |
Yes | |
Analytical EC Column |