missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ NucleoSpin™ 96 Blood QuickPure, 96-Well Kit

Ultra-fast isolation of highly concentrated DNA from blood in proven 96‐well plate format.

Brand:  Macherey-Nagel Bioanalysis™ 740667.24


 View more versions of this product

Product Code. 12768522

  • 5545.00 € / 2304 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

NucleoSpin™ Blood QuickPure binding plates are to be used in a centrifuge with deep buckets allowing processing of NucleoSpin™ 96 well plates stacked on 96 well blocks. The method is based on a specially treated silica membrane which speeds up the procedure by a combined washing/drying step. The number of washing and drying steps is reduced from 3 to 1. Therefore the hands-on time is minimised. Lysis is achieved by incubation of whole blood in a solution containing large amounts of chaotropic ions in the presence of proteinase K. Appropriate conditions for binding of DNA to the silica membrane of the NucleoSpin™ Blood QuickPure binding strips are created by addition of ethanol to the lysate. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer and can be used directly for PCR, Southern blotting or any kind of enzymatic reaction. The kit allows isolation of 4 μg to 6 μg of pure genomic DNA from 200 μL whole blood with an A260/280 ratio between 1.60 and 1.90 and a typical concentration of 40 to 60 ng/μL.Kit components: NucleoSpin™ Blood QuickPure binding plates, buffers, proteinase K, accessories.

  • Silica membrane technology
  • Manual processing
  • Yield: 4 to 6 μg
  • Fragment size: 300 bp to 30 kb
  • Sample material: human blood or other body fluid (200 μL/300 μL), 107 lymphocytes
  • Elution volume: 75 to 100 μL
  • Binding capacity: 60 μg
  • Preparation time: 30 min/prep
Specifications

Specifications

18°C to 25°C
Isolation of DNA
DNA
96-well Plate
2304 Preps
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Category
  • Skin sensitiser Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers