Learn More
Macherey-Nagel™ NucleoSpin™ Plant II Buffers
Supplemental buffer solutions (lysis, precipitation, elution, binding and wash buffers) for use with NucleoSpin™ Plant II mini, midi or maxi kits for the rapid isolation of genomic DNA from plant samples, with higher yield and quality.
Brand:  Macherey-Nagel™ 740351.1000
 View more versions of this product
Product Code. 15829077
  • 241.00 € / 1000mL
Explore more special offers
Description
Description
NucleoSpin™ Plant II Kits (mini, midi or maxi) are used for rapid preparation of genomic DNA from plant cells and fungi. Kits are available in capacities to accommodate small, medium and large scale processing.
  • Two alternative lysis buffers included for optimal processing of various samples and highest flexibility:
    • Lysis Buffer PL1, based on CTAB lysis method
    • Lysis Buffer PL2, based on SDS lysis method
  • NucleoSpin™ filters included for clarification of lysate
  • Improved buffer compositions – higher yield and purity
  • Optimized silica membrane – improved DNA binding
  • RNase A included
  • Typical downstream applications: PCR, Southern blotting, enzymatic reactions
Specifications
Specifications
Colorless
Dodecyl sulfate, sodium salt; Sodium chloride
1000 mL
Odorless
Liquid
Lysis Buffer PL2
NucleoSpin Plant II kit
7.5 to 8.5
50-01-1
Room Temperature (18°C to 25°C)
Preparation of genomic DNA from plant cells and fungi