missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Plant II Buffers
Supplemental buffer solutions (lysis, precipitation, elution, binding and wash buffers) for use with NucleoSpin™ Plant II mini, midi or maxi kits for the rapid isolation of genomic DNA from plant samples, with higher yield and quality.
69.65 € - 344.00 €
Specifications
For Use With (Equipment) | NucleoSpin Plant II kit |
---|---|
CAS | 50-01-1 |
Content And Storage | Room Temperature (18°C to 25°C) |
Form | Liquid |
For Use With (Application) | Preparation of genomic DNA from plant cells and fungi |
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
15800968
|
Macherey-Nagel™
740937.1 |
Binding Buffer PC | 1000 mL |
311.00 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15819077
|
Macherey-Nagel™
740350.1 |
Elution Buffer PE | 1000 mL |
69.65 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15890958
|
Macherey-Nagel™
740918.1 |
Lysis Buffer PL1 | 1000 mL |
274.00 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15829077
|
Macherey-Nagel™
740351.1000 |
Lysis Buffer PL2 | 1000 mL |
241.00 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15839077
|
Macherey-Nagel™
740352.1000 |
Precipitation Buffer PL3 | 1000 mL |
144.00 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15810968
|
Macherey-Nagel™
740938.1 |
Wash Buffer PW1 | 1000 mL |
344.00 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15820968
|
Macherey-Nagel™
740939.1 |
Wash Buffer PW2 Concentrate | 200 mL |
93.05 €
200mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NucleoSpin™ Plant II Kits (mini, midi or maxi) are used for rapid preparation of genomic DNA from plant cells and fungi. Kits are available in capacities to accommodate small, medium and large scale processing.
- Two alternative lysis buffers included for optimal processing of various samples and highest flexibility:
- Lysis Buffer PL1, based on CTAB lysis method
- Lysis Buffer PL2, based on SDS lysis method
- NucleoSpin™ filters included for clarification of lysate
- Improved buffer compositions – higher yield and purity
- Optimized silica membrane – improved DNA binding
- RNase A included
- Typical downstream applications: PCR, Southern blotting, enzymatic reactions
Specifications
NucleoSpin Plant II kit | |
Room Temperature (18°C to 25°C) | |
Preparation of genomic DNA from plant cells and fungi |
50-01-1 | |
Liquid |
Safety and Handling
missing translation for 'shelfLife' : Up to 1 year