missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plant II, Mini Kit for DNA from Plants

Isolate and purify genomic DNA from plant samples. NucleoSpin™ Plant II kits enable enhanced yield and quality DNA.

Brand:  Macherey-Nagel™ 740770.250

773.67 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



 View more versions of this product

Product Code. 11912262

  • 1010.00 € / 250 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Features
  • The second generation for rapid isolation of genomic DNA from plant samples
  • with higher yield and quality
  • Two alternative lysis buffers included for optimal processing of various samples and highest flexibility:
  • Lysis Buffer PL1, based on CTAB lysis method
  • Lysis Buffer PL2, based on SDS lysis method
  • NucleoSpin™ Filters included for clarification of lysate
  • Improved buffer compositions – higher yield and purity
  • Optimized silica membrane – improved DNA binding
  • RNase A included
Applications
  • DNA from plant cells and tissue
  • DNA from fungi
  • Typical downstream applications: PCR, Southern blotting, enzymatic reactions
Specifications

Specifications

250 Preps
Plant II
<100 mg wet weight, <20 mg dry weight
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride, ethanol/dodecylsulfate, sodium salt/guanidine hydrochloride, 2-propanol

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.