missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ RNA Virus
Complete kit with silica membrane mini spin columns for reliable purification of viral RNA from cell-free biological fluids (e.g., serum and plasma).
Brand: Macherey-Nagel™ 740956.10
This item is not returnable.
View return policy
Description
NucleoSpin™ RNA Virus
- Reliable and reproducible isolation of viral RNA free of impurities
- Safe and well-established procedure
- Proven for a large variety of viruses including influenza viruses, HCV, HIV, Bird Flu Virus, Blue Tongue Virus
- Carrier RNA included for highest sensitivity in downstream applications
- Fast and convenient protocol – viral RNA and DNA* within only 30 min
- Support protocol for parallel isolation of viral RNA and DNA (with Proteinase K, not included in Kit)
- Viral RNA from 150 μL serum, plasma, cell-free biological fluids
- Typical downstream applications: RT-PCR, enzymatic reactions
Specifications
Blue band | |
Mini Spin Column | |
50 μL | |
Binding capacity: 40 μg | |
Isolation of viral RNA | |
10 Preps | |
30 min/4-6 preps |
Room temperature (18 - 25 °C) | |
Silica Membrane | |
Cell-free Biological Fluids, Plasma, Serum | |
Centrifuge | |
NucleoSpin RNA Virus Columns with Collection Tubes, Collection Tubes (2 mL), buffers, Carrier RNA | |
< 150 μL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- NucleoSpin RNA Virus
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash with water thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : guanidine hydrochloride, ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction