missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ RNA Virus
Complete kit with silica membrane mini spin columns for reliable purification of viral RNA from cell-free biological fluids (e.g., serum and plasma).
49.90 € - 1010.00 €
Specifications
Color | Blue band |
---|---|
Content And Storage | Room temperature (18 - 25 °C) |
Format | Mini Spin Column |
Isolation Technology | Silica Membrane |
Elution Volume | 50 μL |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
12771021
|
Macherey-Nagel™
740956.10 |
10 Preps |
49.90 €
10 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12781021
|
Macherey-Nagel™
740956.250 |
250 Preps |
1010.00 €
450mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NucleoSpin™ RNA Virus
- Reliable and reproducible isolation of viral RNA free of impurities
- Safe and well-established procedure
- Proven for a large variety of viruses including influenza viruses, HCV, HIV, Bird Flu Virus, Blue Tongue Virus
- Carrier RNA included for highest sensitivity in downstream applications
- Fast and convenient protocol – viral RNA and DNA* within only 30 min
- Support protocol for parallel isolation of viral RNA and DNA (with Proteinase K, not included in Kit)
- Viral RNA from 150 μL serum, plasma, cell-free biological fluids
- Typical downstream applications: RT-PCR, enzymatic reactions
Specifications
Blue band | |
Mini Spin Column | |
50 μL | |
Binding capacity: 40 μg | |
Isolation of viral RNA | |
< 150 μL |
Room temperature (18 - 25 °C) | |
Silica Membrane | |
Cell-free Biological Fluids, Plasma, Serum | |
Centrifuge | |
NucleoSpin RNA Virus Columns with Collection Tubes, Collection Tubes (2 mL), buffers, Carrier RNA | |
30 min/4-6 preps |
Safety and Handling
missing translation for 'shelfLife' : Up to 1 year
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- NucleoSpin RNA Virus
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash with water thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : guanidine hydrochloride, ethanol