Learn More
Syngene NuGenius XE
GREENER_CHOICE
NuGenius XE offers flexible DNA, stain free, autorads analysis. Intuitive touch screen system. Compact benchtop size with built in PC.
Brand:  Syngene NuGeniusXE
Product Code. 17922952
  • 10655.00 € / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • NuGenius XE offers flexible DNA, stain free, autorads analysis.
  • Intuitive touch screen system.
  • Compact benchtop size with built in PC.
  • Internal lighting includes a transilluminator option for working with DNA gels (Ethidium bromide).
  • Our new UV-blue light converter screen allows imaging of a wide range of safe dyes.
  • A visible converter option can quickly extend its use for working with visible gels and blots (Coomassie blue and Silver stain).
  • The overhead LED white lighting is included as standard for easy sample positioning and focussing.
  • Safely excise your DNA bands from an agarose gel with the darkroom door open using the optional safety shield protecting the user when working with UV.
  • With a 5M pixel camera and f/1.2 motor driven lens provides exquisite pixel resolution and unrivalled sensitivity for gels and blots with a viewing area of 20 x 24 cm.
  • Images are saved directly to USB or network for easy transfer.
  • Quickly generate quantitative data with GeneTools analysis software.
  • NuGenius XE comes with unlimited copies of GeneTools.
Specifications
Specifications
Touchscreen
5 MP
Gel Documentation
Gel Imaging System
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Syngene NuGenius XE > Display: Touchscreen