Learn More
Tocris Bioscience™ Selinexor
Selective exportin 1 (XPO1/CRM1) inhibitor
Brand:  Tocris Bioscience™ 7760/10
 View more versions of this product
Product Code. 18737176
  • 211.00 € / 10mg
Explore more special offers
Description
Description
Selinexor is an inhibitor of the exportin 1 (XPO1/CRM1) nuclear exporter. Shows selectivity for TNBC over ER-positive breast cancer cell lines (IC50 values are 11-550 nM and >1000 nM, respectively), and reduces growth of xenografts. Inhibits growth of thyroid cancer cell lines (IC50 = 150-500 nM), potentiates the anti-tumor effects of Olaparib (Cat. No. 7579) in ovarian cancer and breast cancer. Also a potent inhibitor of SARS-CoV-2 replication in vitro (EC50 = 10 nM) and downregulates the production of pro-inflammatory cytokines including TNF-α and IL-6.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
(2Z)-3-[3-[3,5-Bis(trifluoromethyl)phenyl]-1H-1,2,4-triazol-1-yl]-2-propenoic acid 2-(2-pyrazinyl)hydrazide
10 mg
C17H11F6N7O
1393477-72-9
Antiviral Compounds
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tocris Bioscience™ Selinexor > 10 mg