missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 2.0mL Internally-Threaded Universal Tubes
GREENER_CHOICE

Experience the best features of our storage tube portfolio: universal cap, universal latch rack and free-standing tube design -- all for either benchtop or automation use.

Brand:  Thermo Scientific™ 374511-BR

 View more versions of this product

Product Code. 15513734

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Natural
Plastic
Universal Tubes in Latch Rack
Internal Thread, Flat Bottom
10 Racks of 48 Tubes
480
White patch and graduations
Nunc
2.0 mL
Sterile SAL 10-6
2D Barcode (Rack Only)
Natural
48
Cryogenic
480
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ 2.0mL Internally-Threaded Universal Tubes > Universal tubes without 2D code in barcoded latch rack

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.