Learn More
Thermo Scientific™ Nunc™ IVF Multi Dish, 4-well

Perform applications which yield consistently reliable, high-quality results during vitrification.
441.00 € - 474.00 €
Specifications
Certifications/Compliance | CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested |
---|---|
Culture Area | 1.9 cm2/well |
Material | Polystyrene (USP Class VI) |
Sterility | Sterile |
Well Count | 4 |
Product Code | Brand | Surface Treatment | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Price | Quantity & Availability | |||||
10404532
![]() |
Thermo Scientific™
144444 |
Nunclon Delta |
474.00 €
Pack of 120 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10756864
|
Thermo Scientific™
179830 |
Non-treated |
441.00 €
Pack of 120 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Generate consistently reliable results during IVF and embryo vitrification with Thermo Scientific™ Nunc™ 4-Well IVF Dishes, which meet the rigorous requirements of IVF testing, certification and validation.
Innovative features for high quality and ease-of-use:
- Ensures proper well orientation and prevents cross contamination with notched lid and dish
- Prevents scratches on well bottoms and promotes fast temperature equilibrium with 0.8mm raised edge
- The Nunclon Delta surface treatment ensures consistency of a more hydrophilic surface
- The non-treated hydrophobic surface facilitates microdrop embryo culture
- Certificates of conformity available for every lot
Rigorously tested, certified and validated for IVF:
- CE-marked (Notified Body 2460), FDA-listed
- Produced under ISO 13485 with full lot traceability
- Sterile (SAL 10-6)
- Non-pyrogenic (LAL tested)
- USP Class VI polystyrene
- Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
Warranty: 90 days
Specifications
CE-marked (Notified Body 2460), FDA-listed, MEA-tested, HSSA-tested | |
Polystyrene (USP Class VI) | |
4 | |
4 | |
66 mm |
1.9 cm2/well | |
Sterile | |
120 | |
66 mm |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.