Learn More
Thermo Scientific™ Nunc™ 96-Well Polypropylene DeepWell™ Sample Processing & Storage Plates with Shared-Wall Technology
Optimize storage capacity and improve mixing using these DeepWell™ storage plates featuring shared-wall technology. Shared-Wall reduces the height of the plate, increases the diameter of the well for sample extraction.
Brand: Thermo Scientific™ 278752
352.91 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
Description
Optimize storage capacity and improve mixing using Thermo Scientific™ Nunc™ 1.3 and 2.0mL DeepWell™ Plates with Shared-Wall Technology. Ideal for sample collection, storage, combinatorial chemistry and library applications, and resistant to most chemicals, solvents and alcohols used in combinatorial chemistry.
The large well diameters offer ideal access and excellent mixing characteristics, and the U shaped well enhances compatibility with the magnetic bead extraction. The V shaped well bottom ensures low dead-volume and high sample recovery during extraction
All Thermo Scientific plates are manufactured in our cleanroom facility or clean environment to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.
Features:
- Ideal as a collection plate for Nunc Filter Plates
- Storage plate for compounds, samples or biomolecules
- Round-bottom wells reduce liquid retention
- Round well enhances mixing
- Automated molecular diagnostic workflows
- Magnetic Bead Extraction compatible
- Ideal for Kitting
Working volumes:
- 260251 and 260252 50-1000 μL/well
- 278743 and 278752 50-1900 μL/well
Recommended for:
Sample collection; storage; combinatorial chemistry; drug library applications; bacterial and yeast growth; collection plates for Nunc filter plates; NGS and genomic library prep
Compatible Sealing: Adhesive and Heat seals
Hazardous: No
Max Centrifugal Force: 2000 g
All plates are ANSI format for compatibility with automated systems.
To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.
Specifications
Custom Barcoding Available | |
96-well | |
96-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Low binding PP | |
Natural | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
2000 μL | |
1700 μL | |
Non-treated | |
Yes | |
60 | |
60/Cs. | |
Yes | |
ANSI/ SLAS Standard |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
Without Lid | |
8 x 12 | |
60/Case | |
Nunc | |
96-well plate | |
Polypropylene | |
96 | |
Non-sterile | |
Round | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
5 | |
For Research Use Only. Not for use in diagnostic procedures. | |
PCR Ready |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.