Learn More
Thermo Scientific™ Nunc™ EasyFill™ Cell Factory™ Systems
Save valuable space with Thermo Scientific™ Nunc™ EasyFill™ Cell Factory system. Each 10-layer, single-use system holds the equivalent of 36 T175 flasks.
Brand: Thermo Scientific™ 140400
296.22 EUR valid until 2024-12-31
Use promo code "21939" to get your promotional price.
Description
Save valuable space with Thermo Scientific™ Nunc™ EasyFill™ Cell Factory system. Each 10-layer, single-use system holds the equivalent of 36 T175 flasks. The linear multi-layer cell culture format is easy to use and promotes easy scalability, ideal for both research and commercial-scale applications.
- "Out of the box" system
- Large, vented screw closure
- Versatile port design for pouring and aseptic filling
- Enhance productivity— fill and empty five times faster than T175 flasks
- Achieve fast start-up—no accessories required with EasyFill Cell Factory systems
Simply pour the media directly into the EasyFill Cell Factory system's large opening.
Specifications
EasyFill Cell Factory | |
1 | |
Sterile | |
Vented | |
10 |
2 | |
Cell Factory System | |
Flat | |
6320 cm² |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.