Learn More
Thermo Scientific™ Nunc™ Lab-Tek™ II Chamber Slide™ System
Grow cells directly on slides with the Thermo Scientific™ Nunc™ Lab-Tek™ II Chamber Slide™ System.
Brand:  Thermo Scientific™ 154534
 View more versions of this product
Product Code. 11397704
Description
Description
Grow, stain, and examine cells directly on the Thermo Scientific™ Nunc™ Lab-Tek™ II Chamber Slide™ which consists of a removable polystyrene media chamber attached to a standard glass slide. Slide surface is specially treated for adherent cell culture. Available in 1-, 2-, 4-, and 8-well formats.
  • Completely sterile and available in 1-, 2-, 4- or 8-well configurations
  • Tabbed cover fits on top of the chamber to allow gas exchange while minimizing evaporation
  • Removal tool (included) makes it easy to separate the chamber from slide
  • Superfrost™ marking area facilitates labeling
  • Non-fluorescent microscope slide, glass with rounded corners
  • Biocompatible adhesive
  • Inert hydrophobic well border printed on slide
  • Treated for excellent attachment and growth of cells
  • Slide separator included in each pack
  • For research use only
Includes:
Removal tool
Note:
Superfrost™ is a registered trademark of Erie Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8
Polystyrene
Cell Culture
0.2 to 0.5 mL
16
Sterile
0.7 cm2
8-well Chamber Slide w/ removable wells
RS Glass
Clear polystyrene lid with handles on the top and side for aseptic manipulations
74 x 25 mm
Clear
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ Lab-Tek™ II Chamber Slide™ System > 8-well; Growth area: 0.7cm2; Working Volume: 0.2 to 0.5mL
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.