missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Microplate Lids
BIOPHARMAGREENER_CHOICE

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.

89.85 € - 581.00 €

Specifications

Material Polystyrene
Length (English) 5 in.
Length (Metric) 127 mm
Width (English) 3.35 in.
Width (Metric) 85 mm
View More Specs

Products 12
Product Code Brand Plate Compatibility Sterility Condensation Rings Cut-off Corners Price Quantity & Availability  
Product Code Brand Plate Compatibility Sterility Condensation Rings Cut-off Corners Price Quantity & Availability  
10392642
GREENER_CHOICE
View Documents
Thermo Scientific™
250002
All Microplates Sterile No No
224.64 €
Pack of 60
In Stock
to see stock.
Edge
Add to basket
 
10046891
GREENER_CHOICE
View Documents
Thermo Scientific™
250003
All Microplates Non-sterile No No
193.00 €
Pack of 60
In Stock
to see stock.
Edge
Add to basket
 
10622212
GREENER_CHOICE
View Documents
Thermo Scientific™
250005
All Microplates Non-sterile No No
581.00 €
Pack of 180
In Stock
to see stock.
Add to basket
 
10265502
GREENER_CHOICE
View Documents
Thermo Scientific™
253623
All Microplates Non-sterile Yes No
480.00 €
Pack of 100
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
 
10275502
GREENER_CHOICE
View Documents
Thermo Scientific™
253624
All Microplates Sterile Yes No
502.00 €
Pack of 100
Estimated Shipment: 24-06-2025
to see stock.
Add to basket
 
10040401
BIOPHARMAGREENER_CHOICE
View Documents
Thermo Scientific™
263339
96-well Plate Non-sterile Yes Long Side
104.00 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
10334311
BIOPHARMA
View Documents
Thermo Scientific™
264122
96-well Plate Sterile Yes Long Side
89.85 €
Pack of 50
In Stock
to see stock.
Edge
Add to basket
 
10752753
GREENER_CHOICE
View Documents
Thermo Scientific™
264611
384-well Plate Sterile No Long Side
329.00 €
Pack of 60
In Stock
to see stock.
Add to basket
 
10393072
GREENER_CHOICE
View Documents
Thermo Scientific™
264612
384-well Plate Non-sterile No Long Side
340.00 €
Pack of 180
In Stock
to see stock.
Add to basket
 
10766723
GREENER_CHOICE
View Documents
Thermo Scientific™
264616
384-well Plate Sterile No Long Side
397.00 €
Pack of 180
Estimated Shipment: 15-08-2025
to see stock.
Add to basket
 
10242913
GREENER_CHOICE
View Documents
Thermo Scientific™
264623
96-well Plate Sterile Yes Long Side
122.00 €
Pack of 60
In Stock
to see stock.
Add to basket
 
10601922
GREENER_CHOICE
View Documents
Thermo Scientific™
448143
384/1536-well Plate, OmniTray Non-sterile No Long Side N/A N/A N/A
Description

Description

Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.

  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
Specifications

Specifications

Polystyrene
127 mm
85 mm
5 in.
3.35 in.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ Microplate Lids

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.