Learn More
Thermo Scientific™ Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Brand: Thermo Scientific™ 264122
63.43 EUR valid until 2025-05-31
Use promo code "25075" to get your promotional price.
Description
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
- Lids are for use with any standard plates
- 96 Well Lids have individual condensation rings
- Lids have notches for quick orientation
- Universal Lids can be used on any standard height plate on the market

Specifications
Clear | |
Polystyrene | |
96 | |
5 in. | |
Yes | |
50 | |
5 x 3.35 in. (127 x 85 mm) | |
3.35 in. | |
Plate Lid | |
None | |
All well shapes | |
Long Side |
5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm) | |
Sterile | |
7.9 mm | |
127 mm | |
0.3 in. | |
1 | |
Yes | |
85 mm | |
Nunc | |
-20°C to +80°C | |
96-well Plate |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
183961 | Certificate of Compliance | 12/12/2024 | 264122 |
172569 | Certificate of Compliance | 03/12/2024 | 264122 |
182569 | Certificate of Compliance | 03/12/2024 | 264122 |
181550 | Certificate of Compliance | 03/12/2024 | 264122 |
183961 | Certificate of Irradiation | 11/12/2024 | 264122 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.