Learn More
Thermo Scientific™ 96 Well White/Clear Bottom Plate, TC Surface
Color:White, Clear Bottom; Sterility:Sterile; Lid:With Lid; Type: Optical Polymer Base
Brand: Thermo Scientific™ 165306
413.40 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Thermo Scientific™ Nunc MicroWell 96-Well Optical-Bottom Plates with Polymer Base have a solid polystyrene black or white upper structures with a polystyrene film at the bottom of the plate. The plates are optimized for fluorescence and luminescence in cell culture and microscopic applications.
White or black polystyrene upper structure with polymer or coverglass bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications.
- Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
- Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
- Available in black for fluorescence assays or white for luminescence and fluorescence assays; both with clear bottoms
- Polymer base plate
- Working range, 50-200μL/well
- Customized bar coding is available
Ordering Information:
Part No. 152040, 152036, 152028 and 152037 is produced in a clean environment.

Specifications
Custom Barcoding Available | |
With Lid | |
10 Pk., 30 Cases | |
White | |
High Flange | |
400 μL | |
50 to 200 μL | |
Cell Culture | |
30 | |
Case of 30 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
8 x 12 | |
Nunc | |
Optical Polymer Base | |
Polystyrene | |
96 | |
Sterile | |
Flat Bottom (F-well) | |
10 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1424617 | Certificate of Processing | 07/04/2025 | 165306 |
1424648 | Certificate of Processing | 04/04/2025 | 165306 |
1424615 | Certificate of Processing | 04/04/2025 | 165306 |
1425854 | Certificate of Compliance | 24/03/2025 | 165306 |
1424649 | Certificate of Compliance | 23/03/2025 | 165306 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.