Learn More
Thermo Scientific™ Nunc™ Petri Dishes
BIOPHARMA
Prepare samples in hospital environments, food science or the pharmaceutical industry with these petri dishes
216.00 € - 221.00 €
Specifications
Bottom Shape Flat
Material Polystyrene
Shape Round
Sterility Sterile
Type Petri Dish
View More Specs
Products 2
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
10051622
BIOPHARMA
View Documents
Thermo Scientific™
249964
140 mm
221.00 €
Pack of 80
In Stock
to see stock.
Edge
Add to basket
10444912
BIOPHARMA
View Documents
Thermo Scientific™
263991
100 mm
216.00 €
Pack of 320
In Stock
to see stock.
Edge
Add to basket
Description
Description
Prepare samples in hospital environments, food science or the pharmaceutical industry with Thermo Scientific™ Nunc™ Petri Dishes. These dishes are available with a molded grid and perform well in automatic dispensers due to complete flatness and uniform height.
  • For the culturing of bacteria, fungi and other microorganisms
  • Constructed of high-quality and optically clear polystyrene, with lid
  • Fitted with top and bottom stacking rings for easier handing and stable stacking
  • Vented for gaseous exchange
  • Sterilized by gamma irradiation
Specifications
Specifications
Flat
Round
Petri Dish
Polystyrene
Sterile
1
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ Petri Dishes