Learn More
Product Code | Brand | Material | For Use With (Application) | Certified RNase/DNase-free | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | For Use With (Application) | Certified RNase/DNase-free | Price | Quantity & Availability | |||||
10716943
![]() |
Thermo Scientific™
232698 |
Aluminum | PCR, storage in 96-well plates | Yes |
121.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
10681864
![]() |
Thermo Scientific™
232699 |
Aluminum | PCR, storage in 384 well plates | Yes |
125.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10265411
![]() |
Thermo Scientific™
232701 |
Polypropylene | PCR | Yes |
202.80 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10194411
![]() |
Thermo Scientific™
232702 |
Polypropylene | PCR, fluorescence assays, microscopy | Yes |
175.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10633973
![]() |
Thermo Scientific™
235306 |
Polyethylene | Low autofluorescence, ELISA | No |
581.00 €
Pack of 100 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10275452
![]() |
Thermo Scientific™
235307 |
Polyolefin | PCR, qPCR, ELISA, Protein crystallography | Yes |
534.06 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11371555
![]() |
Thermo Scientific™
236272 |
Vinyl | Luminescence, photosensitive samples | No |
68.80 €
Pack of 50 |
Estimated Shipment: 07-04-2025
Log in to see stock. |
|||||
10170671
![]() |
Thermo Scientific™
236366 |
Polyester | Cell culture, sterile applications | No |
637.00 €
Pack of 800 |
In Stock
Log in to see stock. ![]() |
|||||
12770006
![]() |
Thermo Scientific™
236366 |
Polyester | Cell culture, sterile applications | No | N/A | ||||||
11948231
![]() |
Thermo Scientific™
236370 |
Polyester | General purpose, sample detection | No |
148.91 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
10160993
![]() |
Thermo Scientific™
236703 |
Vinyl | Fluorescence, photosensitive samples | No |
103.95 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
10552615
![]() |
Thermo Scientific™
236707 |
Polyester | General purpose, ELISA | Yes |
662.00 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
10681344
![]() |
Thermo Scientific™
241205 |
Non-woven Rayon | Breathable, Cell culture | No |
194.00 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
10082182
![]() |
Thermo Scientific™
249720 |
Non-woven Rayon | Cell growth, Bacterial growth | Yes |
162.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
10777042
![]() |
Thermo Scientific™
250050 |
Plastic | Consistent seal | No |
61.30 €
Pack of 2 |
In Stock
Log in to see stock. |
|||||
10574561
![]() |
Thermo Scientific™
276014 |
Aluminum | PCR, long-term storage | Yes |
198.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
10333925
![]() |
Thermo Scientific™
3501 |
Polyester | Short-term storage, ELISA | No |
97.05 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
10304315
![]() |
Thermo Scientific™
5701 |
Polyester | Short-term storage, ELISA, minimizes evaporation and cross contamination | No |
85.45 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
10642674
|
Thermo Scientific™
AB-0718 |
Polypropylene | Ventilating and storing cell cultures | No |
144.00 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
11524794
|
Thermo Scientific™
AB-0580 |
PVC-coated Polypropylene | Crystallography and genotyping | No |
68.59 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
Descrizione
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
- Adhesive seals effectively seal all microplate formats
- Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils