missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Serological Pipettes

A full range of quality serological pipettes to support each stage of cell culture research

Brand:  Thermo Scientific™ 170375N

97.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 16460911

  • 129.00 € / Pack of 200
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Yes
Serological Pipette, 5 year shelf life
Sterile
Bulk, 25/bag
Rnase and DNase free, human DNA free, non-cytotoxic, non-pyrogenic
Polystyrene
25/Pk., 200/Cs.
25 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberBB03017 Certificate TypeCertificate of Analysis Date17/05/2022 Catalog Number170375N
Lot NumberGL17012 Certificate TypeCertificate of Analysis Date11/11/2021 Catalog Number170375N
Lot NumberBA03017 Certificate TypeCertificate of Analysis Date11/11/2021 Catalog Number170375N
Lot NumberGJ17010 Certificate TypeCertificate of Analysis Date11/11/2021 Catalog Number170375N
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ Serological Pipettes >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.