Learn More
Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes
GREENER_CHOICE
Store tubes for automated picking/placing, liquid handling and other high throughput applications using Nunc Universal Latch Rack for Cryogenic Tubes.
Brand:  Thermo Scientific™ 331827
171.94 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 12617316
  • 191.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes. Features a transparent cover with options for tubes of several sizes.
  • Non-sterile
Ordering Information:
Case of 10
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transparent
1 to 2 mL Cryogenic Tubes
Cryogenic Tube Storage Rack (w/Latch)
For 1-2mL Standard Cryotubes, non-sterile
10/pk., 10/cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes > For 1-2mL Standard Cryotubes, non-sterile