Learn More
Brady™ Nylon Cloth Labels for M710
Label Printer Labels for use with Brady™ Printer M710.
Brand:  Brady™ 311987
 View more versions of this product
Product Code. 17897934
  • 83.80 € / Pack of 750
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Labels for the M710 Label Printer
  • This material is UL approved with its respective ribbon
  • Matte, white, polyamide-coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
  • Service temperature of -40°C to +90°C
Specifications
Specifications
Halogen-free (DIN VDE 0472 Part 815)
None
Matte
Nylon
Thermal Transfer
Non-sterile
M710, BMP71
-40°C
Rectangle
-40°C to +90°C
0.166 mm
0.354 in.
15 mm
Permanent Acrylic
Yellow
Label
Single-side Printable
Labels
Component and Equipment Labeling, Electrical Labeling, General Identification, Laboratory Labeling, Warehouse Marking, Wire and Cable Labeling
1 Label
750 Labels
-40°F to +194°F
0.0065 in.
9 x 15 mm
0.59 in.
15 mm